accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHH2C

CAS RN 4516-90-9

IUPAC Name: 2-methylbut-3-en-1-ol
Synonyms: 2-methylbut-3-en-1-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: NVGOATMUHKIQQG-UHFFFAOYNA-N
SMILES: CC(CO)C=C
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)