accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OHONCH3OOCH3CH3H3C

CAS RN 45170-31-8

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)(methyl)amino]-3-methylbutanoic acid|BOC-N-ME-VAL-OH
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: XPUAXAVJMJDPDH-QMMMGPOBSA-N
SMILES: CC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)