CAS RN 4518-98-3
CAS RN 4518-98-3
IUPAC Name:
1,1,2,2-tetrachloro-1,2-dimethyldisilane
Synonyms:
1,1,2,2-tetrachloro-1,2-dimethyldisilane
Molecular Weight (g/mol):
228.04
Molecular Formula:
C2H6Cl4Si2
InChi Key:
JTBAMRDUGCDKMS-UHFFFAOYSA-N
SMILES:
C[Si](Cl)(Cl)[Si](C)(Cl)Cl
1
–
1
of
1
results
1
1,1,2,2-Tetrachloro-1,2-dimethyldisilane 98.0+%, TCI America™
CAS: 4518-98-3 Molecular Formula: C2H6Cl4Si2 Molecular Weight (g/mol): 228.04 MDL Number: MFCD00192469 InChI Key: JTBAMRDUGCDKMS-UHFFFAOYSA-N PubChem CID: 78276 IUPAC Name: 1,1,2,2-tetrachloro-1,2-dimethyldisilane SMILES: C[Si](Cl)(Cl)[Si](C)(Cl)Cl