accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 452-06-2

IUPAC Name: 7H-purin-2-amine
Synonyms: 2-aminopurine
Molecular Weight (g/mol): 135.13
Molecular Formula: C5H5N5
InChi Key: MWBWWFOAEOYUST-UHFFFAOYSA-N
SMILES: NC1=NC=C2NC=NC2=N1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)