accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IF

CAS RN 452-68-6

IUPAC Name: 1-fluoro-4-iodo-2-methylbenzene
Synonyms: 1-fluoro-4-iodo-2-methylbenzene
Molecular Weight (g/mol): 236.03
Molecular Formula: C7H6FI
InChi Key: DYQIYXDSKUUZRI-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CC=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results