accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 452-70-0

IUPAC Name: 4-fluoro-3-methylphenol
Synonyms: 4-fluoro-3-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: RVYGYYVGWSCWGY-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results