accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 452-72-2

IUPAC Name: 4-fluoro-2-methylphenol
Synonyms: 4-fluoro-2-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: GKQDDKKGDIVDAG-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)