accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4521-28-2

IUPAC Name: 4-(4-methoxyphenyl)butanoic acid
Synonyms: P-methoxy-G-phenylbutyric acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: LZHMNCJMXQKSBY-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCCC(O)=O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)