accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4525-65-9

IUPAC Name: 2,4,6-trichlorophenyl formate
Synonyms: 2,4,6-trichlorophenyl formate
Molecular Weight (g/mol): 225.45
Molecular Formula: C7H3Cl3O2
InChi Key: YSUDXADDVAYVNS-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(OC=O)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results