accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 452972-11-1

IUPAC Name: 2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 239.51
Molecular Formula: C11H15BClNO2
InChi Key: XFZFMAUUZHBQSS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results