accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOF

CAS RN 453-14-5

IUPAC Name: 1,3-difluoropropan-2-one
Synonyms: 1,3-difluoropropan-2-one
Molecular Weight (g/mol): 94.06
Molecular Formula: C3H4F2O
InChi Key: HKIPCXRNASWFRU-UHFFFAOYSA-N
SMILES: FCC(=O)CF
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results