accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)HOO

CAS RN 453-17-8

IUPAC Name: (2R)-2,3-dihydroxypropanal
Synonyms: triose D-(+)-glyceraldehyde D-glyceraldehyde
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: MNQZXJOMYWMBOU-VKHMYHEASA-N
SMILES: OC[C@@H](O)C=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results