accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONON

CAS RN 4533-42-0

IUPAC Name: 1-(4-nitrophenyl)-1H-pyrrole
Synonyms: 1-(4-nitrophenyl)pyrrole
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: PWCFKNYSCGRNRW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1C=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results