accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHOOHOOH

CAS RN 4534-68-3

IUPAC Name: butane-1,2,3,4-tetracarboxylic acid
Synonyms: butane-1,2,3,4-tetracarboxylic acid
Molecular Weight (g/mol): 234.16
Molecular Formula: C8H10O8
InChi Key: GGAUUQHSCNMCAU-UHFFFAOYNA-N
SMILES: OC(=O)CC(C(CC(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results