accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 45347-82-8

IUPAC Name: azetidin-3-ol
Synonyms: azetidin-3-ol
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: GMWFCJXSQQHBPI-UHFFFAOYSA-N
SMILES: OC1CNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results