accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 45361-12-4

IUPAC Name: pyrrolidine hydroiodide
Synonyms: pyrrolidine hydroiodide
Molecular Weight (g/mol): 199.04
Molecular Formula: C4H10IN
InChi Key: DMFMZFFIQRMJQZ-UHFFFAOYSA-N
SMILES: I.C1CCNC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results