accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4537-73-9

IUPAC Name: pyrimidine-2-carbothioamide
Synonyms: pyrimidine-2-thiocarboxamide
Molecular Weight (g/mol): 139.18
Molecular Formula: C5H5N3S
InChi Key: QAUHVPUYFSGVME-UHFFFAOYSA-N
SMILES: NC(=S)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results