accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4538-37-8

IUPAC Name: 1,4-diisocyanatobutane
Synonyms: 1,4-diisocyanatobutane
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: OVBFMUAFNIIQAL-UHFFFAOYSA-N
SMILES: O=C=NCCCCN=C=O
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)