accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFF

CAS RN 454-31-9

IUPAC Name: ethyl 2,2-difluoroacetate
Synonyms: ethyl 2,2-difluoroacetate
Molecular Weight (g/mol): 124.09
Molecular Formula: C4H6F2O2
InChi Key: GZKHDVAKKLTJPO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)F
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)