accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 454-91-1

IUPAC Name: 1-[3-(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 1-[3-(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 190.17
Molecular Formula: C9H9F3O
InChi Key: YNVXCOKNHXMBQC-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)