accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO

CAS RN 4541-32-6

IUPAC Name: 2,2-dimethylcyclopentan-1-one
Synonyms: 2,2-dimethylcyclopentan-1-one
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: FTGZMZBYOHMEPS-UHFFFAOYSA-N
SMILES: CC1(C)CCCC1=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)