accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4545-21-5

CH3SOOHNNH3C(E)

CAS RN 4545-21-5

IUPAC Name: 4-methyl-N'-[(1E)-1-phenylethylidene]benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-[(1E)-1-phenylethylidene]benzenesulfonohydrazide
Molecular Weight (g/mol): 288.37
Molecular Formula: C15H16N2O2S
InChi Key: MIXFDFCKBMCLGN-DTQAZKPQSA-N
SMILES: C\C(=N/NS(=O)(=O)C1=CC=C(C)C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results