accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 4547-57-3

IUPAC Name: 2-(4-butoxyphenyl)acetic acid
Synonyms: acetic acid, (p-butoxyphenyl)-
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: KLJMYYFCWBVKEE-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(CC(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results