accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 4549-33-1

IUPAC Name: 1,9-dibromononane
Synonyms: 1,9-dibromononane
Molecular Weight (g/mol): 286.05
Molecular Formula: C9H18Br2
InChi Key: WGAXVZXBFBHLMC-UHFFFAOYSA-N
SMILES: BrCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (4)