accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFF

CAS RN 455-01-6

IUPAC Name: 2-[3-(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 2-[3-(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 190.17
Molecular Formula: C9H9F3O
InChi Key: YDKIPCCKZKQMDT-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results