accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 455-18-5

IUPAC Name: 4-(trifluoromethyl)benzonitrile
Synonyms: 4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H4F3N
InChi Key: DRNJIKRLQJRKMM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)