accessibility menu, dialog, popup

UserName

Viewing profile as user:
FF

CAS RN 455-31-2

IUPAC Name: (difluoromethyl)benzene
Synonyms: (difluoromethyl)benzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: JDZLOJYSBBLXQD-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results