accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFF

CAS RN 455-86-7

IUPAC Name: 3,4-difluorobenzoic acid
Synonyms: 3,4-fifluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: FPENCTDAQQQKNY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)