accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 455-93-6

IUPAC Name: 2-fluoro-1-methoxy-4-nitrobenzene
Synonyms: 2-fluoro-1-methoxy-4-nitrobenzene
Molecular Weight (g/mol): 171.13
Molecular Formula: C7H6FNO3
InChi Key: XGMVTXUXZUPGGY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results