accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOO

CAS RN 4551-69-3

IUPAC Name: 4-benzoyl-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 4-benzoyl-5-methyl-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 278.31
Molecular Formula: C17H14N2O2
InChi Key: JDOLPAMOKSAEMD-UHFFFAOYNA-N
SMILES: CC1=NN(C(=O)C1C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)