accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 4553-62-2

IUPAC Name: 2-methylpentanedinitrile
Synonyms: 2-methylpentanedinitrile
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: FPPLREPCQJZDAQ-UHFFFAOYNA-N
SMILES: CC(CCC#N)C#N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)