accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHHNOHSOOHO

CAS RN 4554-26-1

IUPAC Name: piperazine; sulfuric acid
Synonyms: piperazine; sulfuric acid
Molecular Weight (g/mol): 184.21
Molecular Formula: C4H12N2O4S
InChi Key: MYNIYCGOBKAQAO-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.C1CNCCN1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results