accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNOF

CAS RN 456-06-4

IUPAC Name: 4-fluorobenzohydrazide
Synonyms: 4-fluorobenzohydrazide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: UIVXXFYJRYVRKJ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results