accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNHF

CAS RN 456-14-4

IUPAC Name: hydrogen 4-fluorobenzene-1-carboximidamide chloride
Synonyms: hydrogen 4-fluorobenzenecarboximidamide chloride
Molecular Weight (g/mol): 174.6
Molecular Formula: C7H8ClFN2
InChi Key: JQDATBKJKUWNGA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)