accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 456-42-8

IUPAC Name: 1-(chloromethyl)-3-fluorobenzene
Synonyms: M-fluorobenzyl chloride
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: XBDXMDVEZLOGMC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CCl)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)