accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NFHOO

CAS RN 456-88-2

IUPAC Name: 2-amino-3-(3-fluorophenyl)propanoic acid
Synonyms: 3-fluorophenylalanine
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: VWHRYODZTDMVSS-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC(F)=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results