CAS RN 4565-31-5
IUPAC Name:
5-formylthiophene-2-carboxylate
Synonyms:
5-formylthiophene-2-carboxylate
Molecular Weight (g/mol):
155.15
Molecular Formula:
C6H3O3S
InChi Key:
VFEAMMGYJFFXKV-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(S1)C=O
1
–
1
of
1
results
1
5-Formyl-2-thiophenecarboxylic Acid 98.0+%, TCI America™
CAS: 4565-31-5 Molecular Formula: C6H3O3S Molecular Weight (g/mol): 155.15 MDL Number: MFCD00191437 InChI Key: VFEAMMGYJFFXKV-UHFFFAOYSA-M Synonym: 5-Formylthenoic Acid PubChem CID: 818882 IUPAC Name: 5-formylthiophene-2-carboxylate SMILES: [O-]C(=O)C1=CC=C(S1)C=O