accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3OHSN

CAS RN 4568-71-2

IUPAC Name: 3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
Synonyms: 3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
Molecular Weight (g/mol): 269.79
Molecular Formula: C13H16ClNOS
InChi Key: IWSVLBKHBJGMAA-UHFFFAOYSA-M
SMILES: [Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (6)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (5)
  • (4)