accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3NHNONH2N(Z)

CAS RN 4569-88-4

IUPAC Name: 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxonaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular Weight (g/mol): 506.01
Molecular Formula: C30H24ClN5O
InChi Key: SCMDRBZEIUMBBQ-UHFFFAOYSA-N
SMILES: [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)