accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNH2

CAS RN 4570-45-0

IUPAC Name: 1,3-oxazol-2-amine
Synonyms: oxazole-2-amine
Molecular Weight (g/mol): 84.08
Molecular Formula: C3H4N2O
InChi Key: ACTKAGSPIFDCMF-UHFFFAOYSA-N
SMILES: NC1=NC=CO1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)