accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3CH3H3C

CAS RN 4574-04-3

IUPAC Name: trimethyl(tetradecyl)azanium chloride
Synonyms: trimethyl(tetradecyl)azanium chloride
Molecular Weight (g/mol): 291.95
Molecular Formula: C17H38ClN
InChi Key: CEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)