accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNHOCl

CAS RN 457618-46-1

IUPAC Name: 4-chloro-N-(2,4-dichlorophenyl)butanamide
Synonyms: 4-chloro-N-(2,4-dichlorophenyl)butanamide
Molecular Weight (g/mol): 266.55
Molecular Formula: C10H10Cl3NO
InChi Key: SPTDUZNBZNYZFE-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results