CAS RN 457961-34-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Opn4 Antagonist
Prevents melanopsin/Opn4 photoactivation in a reversible manner. Inhibits cellular phototransduction mediated by human Opn4 (IC50 = 665 nM; using CHO transfectants).