CAS RN 458-05-9
IUPAC Name:
(3-fluorophenyl)thiourea
Synonyms:
3-fluorophenylthiourea
Molecular Weight (g/mol):
170.21
Molecular Formula:
C7H7FN2S
InChi Key:
YKOBLHQALWQKTC-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=CC=CC(F)=C1
1
–
1
of
1
results
1
Promotions Available
1-(3-Fluorophenyl)-2-thiourea, 98%, Thermo Scientific™
1-(3-Fluorophenyl)-2-thiourea, 98%, Thermo Scientific™
CAS: 458-05-9 Molecular Formula: C7H7FN2S Molecular Weight (g/mol): 170.21 InChI Key: YKOBLHQALWQKTC-UHFFFAOYSA-N Synonym: 1-3-fluorophenyl-2-thiourea,1-3-fluorophenyl thiourea,3-fluorophenyl thiourea,n-3-fluorophenyl thiourea,amino 3-fluorophenyl amino methane-1-thione,acmc-20apbn,1-m-fluorophenyl thiourea,3-fluoro-phenyl-thiourea PubChem CID: 2734799 IUPAC Name: (3-fluorophenyl)thiourea SMILES: C1=CC(=CC(=C1)F)NC(=S)N