accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 458-50-4

IUPAC Name: 1-bromo-2-fluoro-4-methoxybenzene
Synonyms: 1-bromo-2-fluoro-4-methoxybenzene
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: XANVIFOBBVAKCY-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results