accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 458532-84-8

IUPAC Name: 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 239.51
Molecular Formula: C11H15BClNO2
InChi Key: UUEQDBHKMOFLDP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=NC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results