accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 458532-94-0

IUPAC Name: (2-bromopyridin-4-yl)boronic acid
Synonyms: 2-bromopyridin-4-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: DFIKFQMGZNTFLI-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Br)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results