accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHNCl

CAS RN 458532-96-2

IUPAC Name: (2-chloropyridin-4-yl)boronic acid
Synonyms: 2-chloropyridin-4-ylboronic acid
Molecular Weight (g/mol): 157.36
Molecular Formula: C5H5BClNO2
InChi Key: WJYRVVDXJMJLTN-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=NC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results