accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 459-32-5

IUPAC Name: (2E)-3-(4-fluorophenyl)prop-2-enoic acid
Synonyms: (2E)-3-(4-fluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 166.15
Molecular Formula: C9H7FO2
InChi Key: ISMMYAZSUSYVQG-ZZXKWVIFSA-N
SMILES: OC(=O)\C=C\C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)