accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 459-80-3

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dienoic acid
Synonyms: geranic acid decaprenoic acid nerolic acid
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: ZHYZQXUYZJNEHD-VQHVLOKHSA-N
SMILES: CC(C)=CCC\C(C)=C\C(O)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)